Order Entry
Sweden
ContactUsLinkComponent
Ochratoxin A Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Ochratoxin A in feed, grain sample. 1 * 48 Tests
Catalog # ANTIA327237-48
Supplier:  ANTIBODIES.COM
New Product
Ochratoxin A Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Ochratoxin A in feed, grain sample. 1 * 48 Tests
Catalog # ANTIA327237-48
Supplier:  ANTIBODIES.COM
New Product