Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
RECOMBINANT HUMAN ACTIVIN E PLUS (AVI-TA (Avi-tag) 1000 µG (Microgram) 1 * 1.000 µG
Catalog # MSPPQK082-1000
Supplier:
MARKET SOURCE PART PROCESS
Expand image
RECOMBINANT HUMAN ACTIVIN E PLUS (AVI-TA (Avi-tag) 1000 µG (Microgram) 1 * 1.000 µG
Catalog # MSPPQK082-1000
Supplier:
MARKET SOURCE PART PROCESS