Order Entry
Sweden
ContactUsLinkComponent
Mouse CBS ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse CBS in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA6153-96
Supplier:  ANTIBODIES.COM
New Product
Mouse CBS ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse CBS in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA6153-96
Supplier:  ANTIBODIES.COM
New Product