Order Entry
Sweden
ContactUsLinkComponent
Human Selenium Binding Protein 1/SBP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of human Selenium Binding Protein 1/SBP in serum, plasma, tissue homogenates, and other biol 1 * 96 Tests
Catalog # ANTIA74535-96
Supplier:  ANTIBODIES.COM
New Product
Human Selenium Binding Protein 1/SBP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of human Selenium Binding Protein 1/SBP in serum, plasma, tissue homogenates, and other biol 1 * 96 Tests
Catalog # ANTIA74535-96
Supplier:  ANTIBODIES.COM
New Product