TriLink® CleanCap® Reagent AG
Catalog # SOLUFN-7113-1MMOL
Supplier: TriLink BioTechnologies
New Product
CAS Number:
Specifications
- Seq:5
- Sample size:1×1 mmol
Specifications
About this item
CleanCap® Reagent AG is TriLink’s patented co-transcriptional capping reagent for in vitro transcription of 5’ capped mRNA resulting in a Cap 1 structure. CleanCap has shown to provide up to 98% capping efficiency.
- Grade: RUO
PLEASE NOTE: CleanCap Reagent AG requires an AG initiator. To learn more, please review the Product Insert.
Caution: Item will be delivered at or below –20 °C and should be stored at or below –20 °C.