Order Entry
Sweden
Orders LinkContactUsLinkComponent
3-Bromophenetole

3-Bromophenetole

Bottle
Catalog # APOSOR322269-500G
Supplier:   Apollo Scientific
3-Bromophenetole
Catalog # APOSOR322269-500G
Supplier:  Apollo Scientific
CAS Number:  2655-84-7
3-Bromophenetole

Bottle

CAS Number:  2655-84-7
Price
13 375,00 kr
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  3-Bromophenyl ethyl ether, 1-Bromo-3-ethoxybenzene
  • CAS Number:2655-84-7
  • Formula:C₈H₉BrO
  • MDL Number:MFCD00274287
  • Molecular Weight:201.06 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.