Order Entry
Sweden
ContactUsLinkComponent
Pre-separator D360 - dust class M + ATEX Technical specifications: Container volume (litres) 13 Weight (kg) 11 Dimensions L x W x H (mm) 390 x 460 x 650 Scope of delivery: - Pre-separator D360 - dust class M + ATEX - Electrically conductive suction h 1 * 1 items
Catalog # HAVE210052363
Supplier:  VWR Collection
New Product
Pre-separator D360 - dust class M + ATEX Technical specifications: Container volume (litres) 13 Weight (kg) 11 Dimensions L x W x H (mm) 390 x 460 x 650 Scope of delivery: - Pre-separator D360 - dust class M + ATEX - Electrically conductive suction h 1 * 1 items
Catalog # HAVE210052363
Supplier:  VWR Collection
New Product