Order Entry
Sweden
ContactUsLinkComponent
Pre-separator D360 - dust class M Technical specifications: Container volume (litres) 13 Weight (kg) 11 Dimensions L x W x H (mm) 390 x 460 x 650 Scope of delivery: - Pre-separator D360 - dust class M - Electrically conductive suction hose, DN 35 x 1 * 1 items
  HAVE210052356
New Product
Pre-separator D360 - dust class M Technical specifications: Container volume (litres) 13 Weight (kg) 11 Dimensions L x W x H (mm) 390 x 460 x 650 Scope of delivery: - Pre-separator D360 - dust class M - Electrically conductive suction hose, DN 35 x 1 * 1 items
  HAVE210052356
New Product