Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
GENE KNOCKOUT KIT V2 MOUSE GENE KNOCKOUT 1 * 1,5 Nmo
Catalog # MSPPOF60012561-2
Supplier:
MARKET SOURCE PART PROCESS
Expand image
GENE KNOCKOUT KIT V2 MOUSE GENE KNOCKOUT 1 * 1,5 Nmo
Catalog # MSPPOF60012561-2
Supplier:
MARKET SOURCE PART PROCESS