Order Entry
Sweden
ContactUsLinkComponent
Zeta potential analyzer for solid samples based on the principle of streaming potential and streaming current measurement, including: - SurPASS 3 instrument with integrated titration unit - automated pressure control for adsorption kinetics measureme 1 * 1 items
Catalog # PAAR20880
Supplier:  Anton Paar
New Product
Zeta potential analyzer for solid samples based on the principle of streaming potential and streaming current measurement, including: - SurPASS 3 instrument with integrated titration unit - automated pressure control for adsorption kinetics measureme 1 * 1 items
Catalog # PAAR20880
Supplier:  Anton Paar
New Product