Order Entry
Sweden
ContactUsLinkComponent
This eco-conscious safety trainer offers a composite toecap, puncture resistant textile midsole and dual density PU/PU outsole for s uperior grip and energy absorption. The upper is made of a seamless, knitted, single piece of textile with a padded c 1 * 1 items
Katalog # PRTWFC06BKB38
Supplier:  Portwest
New Product
This eco-conscious safety trainer offers a composite toecap, puncture resistant textile midsole and dual density PU/PU outsole for s uperior grip and energy absorption. The upper is made of a seamless, knitted, single piece of textile with a padded c 1 * 1 items
Katalog # PRTWFC06BKB38
Supplier:  Portwest