Order Entry
Sweden
ContactUsLinkComponent
LCJET DRYING OPTION FOR DRYING COLLECTED FRACTIONS AFTER FRACTION COLLECTION WITH TEMPERATURE CONTROL FROM ROOM TEMPERATURE TO 40 C. THIS OPTION IS IDEAL FOR APPLICATION WHERE COLLECTED FRACTIONS NEED TO BE DRIED 1 * 1 Items
  MSPP400-1010-DRY
LCJET DRYING OPTION FOR DRYING COLLECTED FRACTIONS AFTER FRACTION COLLECTION WITH TEMPERATURE CONTROL FROM ROOM TEMPERATURE TO 40 C. THIS OPTION IS IDEAL FOR APPLICATION WHERE COLLECTED FRACTIONS NEED TO BE DRIED 1 * 1 Items
  MSPP400-1010-DRY