0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
WELL PLATE HOLDER TYPE LOW DESIGNED FOR THE MANUAL INSERTION OF WELL PLATES MAXIMUM WELL PLATE HIGH 49 MM 1 * 1 items
Catalog # MSPP19946
Supplier:
MARKET SOURCE PART PROCESS
Expand image
WELL PLATE HOLDER TYPE LOW DESIGNED FOR THE MANUAL INSERTION OF WELL PLATES MAXIMUM WELL PLATE HIGH 49 MM 1 * 1 items
Catalog # MSPP19946
Supplier:
MARKET SOURCE PART PROCESS