Order Entry
Sweden
ContactUsLinkComponent
,,1 component: Hg 0.5mg/l in HNO3 2% ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 8 months.,, ,,Store under normal laboratory conditions, at temperatures between 15C to 25C,, 1 * 100 mL
Catalog # CMPLRM059258L1
New Product
,,1 component: Hg 0.5mg/l in HNO3 2% ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 8 months.,, ,,Store under normal laboratory conditions, at temperatures between 15C to 25C,, 1 * 100 mL
Catalog # CMPLRM059258L1