Order Entry
Sweden
Orders LinkContactUsLinkComponent
Reservoir Microplates

Reservoir Microplates

Reservoir, single cavity, PP, 8 row base geometry, 290 ml, White, 44 mm, -, —
Catalog # AGLS201252-100
Supplier:   AGILENT
Reservoir Microplates
Catalog # AGLS201252-100
Supplier:  AGILENT
Reservoir Microplates
Reservoir Microplates

Reservoir, single cavity, PP, 8 row base geometry, 290 ml, White, 44 mm, -, —

Price
Inquire For Price
Log in for availability
1 * 25 Items
Product Details & Documents

Polypropylene reservoir microplates for automated liquid handlers and handheld pipettors.

  • Large range of cavity volumes, from 2,6 to 300 ml with specialised bottom geometries to optimise access and minimise dead volumes
  • Low binding surfaces for minimal losses of proteins, DNA/RNA, and other biologicals
  • Molded from chemical-resistant polymers, for maintained performance following prolonged exposure to aggressive solutions and solvents
  • Produced to ANSI/SLAS standards, for compatibility with popular labware and devices
  • Compatible with all major platforms and benchtop devices, making these reservoirs automation friendly
  • Low temperature tolerant, suitable for heating or cooling during use
  • Autoclavable, can be sterilised at 121 °C, 1,03 bar for 20 minutes
  • Fully customisable to fit your requirements
  • Manufactured in a DNAse/RNase-free ISO 9001 regulated facility

Agilent polypropylene reservoir microplates are available for a wide range of automated liquid handlers and handheld pipettors. These reservoir microplates are molded using high-quality, chemical-resistant polymers to ANSI/SLAS standard footprints, with optimised base geometries to minimise dead volumes. They are available with 1 to 24 cavities per reservoir plate, container heights between 19 and 44 mm, and with clear or black lids.

These reservoirs are recommended for both method scale-up and scale-down applications as well as for creating design of experiments (DoE) arrays.

Specifications
  • Packed:
  • Sterile:
    -
  • Colour:
    White
  • Description:
    Reservoir, single cavity, PP, 8 row base geometry, 290 ml
  • Height:
    44 mm
  • Seq:
    08