MCA225S-3S00-D QP99 HWL. Scale div. (d) 0.01 mg. max. load 220 g. Draft Shield D & Activated Ionizer and Draft Shield Motors 1 * 1 items
Catalog # 611-5829
Supplier: Sartorius Balances
New Product
Related Information
Cubis® II Modular Premium Balances
Additional Product Details
Cubis® II Ultra-High Resolution Semi-Micro Balances