Order Entry
Sweden
ContactUsLinkComponent
32 4K-MONITOR 32 4K MONITOR, SCREEN RESOLUTION 3840 X 2160, IMAGE FORMAT 16:9, POWER SUPPLY 100 – 240 VAC, 50/60 HZ, VESA 100 AND VESA 200 MOUNTS VIDEO INPUTS: 1X DISPLAY PORT 1.2 1X DVI-D 1X 12G-SDI 1X HDMI 2.0 1X 3G-SDI VIDEO OUTPUTS 1 * 1 Items
Catalog # MSPPTM343
32 4K-MONITOR 32 4K MONITOR, SCREEN RESOLUTION 3840 X 2160, IMAGE FORMAT 16:9, POWER SUPPLY 100 – 240 VAC, 50/60 HZ, VESA 100 AND VESA 200 MOUNTS VIDEO INPUTS: 1X DISPLAY PORT 1.2 1X DVI-D 1X 12G-SDI 1X HDMI 2.0 1X 3G-SDI VIDEO OUTPUTS 1 * 1 Items
Catalog # MSPPTM343
Frequently Bought Together