Order Entry
Sweden
Orders LinkContactUsLinkComponent
Human Recombinant IL-21, ACF

Human Recombinant IL-21, ACF

100 µG
Catalog # STMC78193.1
Marketsource
Human Recombinant IL-21, ACF
Catalog # STMC78193.1
Marketsource
CAS Number:  
Human Recombinant IL-21, ACF
Human Recombinant IL-21, ACF

100 µG

CAS Number:  
Price
11 541,00 kr
Log in for availability
1 * 100 µG
Product Details & Documents

Interleukin 21 (IL-21) is a pleiotropic cytokine that is composed of four α-helical bundles and primarily produced by natural killer T (NKT) cells, T follicular helper (Tfh) cells, and Th17 cells (Spolski and Leonard 2008). IL-21 signals via heterodimers of the IL-21 receptor (IL-21R) and the IL2RG-encoded common cytokine receptor γ-chain (Parrish-Novak et al.; Ozaki K et al. 2000), and utilizes the JAK/STAT, MAPK, and PI3K pathways (Spolski and Leonard 2014). IL-21 has been shown to have a critical role in regulating immunoglobulin production and differentiation of the pro-inflammatory Th17 population of cells (Ozaki et al. 2002; Nurieva et al.). Additionally, IL-21 specifically sustains CD8+ T cell effector activity and provides a mechanism of CD4+ T cell help during chronic viral infection (Elsaesser et al.). IL-21 signaling was also found critical for the development of type 1 diabetes in NOD mice (Sutherland et al.) and control of T cell autoimmunity by regulatory B cells (Yoshizaki et al.). This product is animal component-free.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications

Specifications

  • Pk:
    100 µG
  • Cat. No.:
    STMC78193.1
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    IL-21, ACF
  • Purity:
    ≥95%
  • Species:
    Human

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.