0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
HX-105, ACTIVE MOBILE PHASE HEATER, 10 X 0.040 , HASTELLOY TUBING, RATED TO 5ML/MIN TO 50ML/MIN @ 60C 1 * 1 Items
Catalog # MSPPHX105-H
Supplier:
MARKET SOURCE PART PROCESS
Expand image
HX-105, ACTIVE MOBILE PHASE HEATER, 10 X 0.040 , HASTELLOY TUBING, RATED TO 5ML/MIN TO 50ML/MIN @ 60C 1 * 1 Items
Catalog # MSPPHX105-H
Supplier:
MARKET SOURCE PART PROCESS