Order Entry
Sweden
ContactUsLinkComponent
1p12 CISH Probe is designed for the qualitative detection of human chromosome 1 specific sequences in formalin-fixed, paraffin-embed ded specimens by chromogenic in situ hybridization (CISH). 1 * 400 µl
Catalog # ABNOCG0035.400UL
Supplier:  Abnova
1p12 CISH Probe is designed for the qualitative detection of human chromosome 1 specific sequences in formalin-fixed, paraffin-embed ded specimens by chromogenic in situ hybridization (CISH). 1 * 400 µl
Catalog # ABNOCG0035.400UL
Supplier:  Abnova