Order Entry
Sweden
ContactUsLinkComponent
CISH Implementation Kit 2 is intended to be used in combination with digoxigenin/dinitrophenyl-labeled CISH probes for the qualitati ve detection of genetic abbreviations, e.g., translocations, deletions, amplifications, and chromosome aneuploidies, 1 * 1 KIT
Catalog # ABNOKA6904.1KIT
Supplier:  Abnova
CISH Implementation Kit 2 is intended to be used in combination with digoxigenin/dinitrophenyl-labeled CISH probes for the qualitati ve detection of genetic abbreviations, e.g., translocations, deletions, amplifications, and chromosome aneuploidies, 1 * 1 KIT
Catalog # ABNOKA6904.1KIT
Supplier:  Abnova