Order Entry
Sweden
ContactUsLinkComponent
LDH-Cytotoxicity Assay Kit is used for rapid quantitative determination of cytotoxicity based on lactate dehydrogenase released into cell culture medium. 1 * 1 KIT
Catalog # ABNOKA6677.1KIT
Supplier:  Abnova
LDH-Cytotoxicity Assay Kit is used for rapid quantitative determination of cytotoxicity based on lactate dehydrogenase released into cell culture medium. 1 * 1 KIT
Catalog # ABNOKA6677.1KIT
Supplier:  Abnova
Frequently Bought Together