0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 32 g. Draft Shield D & Activated Draft Shield Motors 1 * 1 items
SARBMCA36S3S004M
:
Sartorius Balances
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 32 g. Draft Shield D & Activated Draft Shield Motors 1 * 1 items
SARBMCA36S3S004M
:
Sartorius Balances