Order Entry
Sweden
Orders LinkContactUsLinkComponent
Double Ended Spoons
Double Ended Spoons
Double ended spoon, 250 mm, 1,8/5 ml, Stainless steel
Katalog # SAMPA104-250
Leverantör:   Sampling Systems
Double Ended Spoons
Katalog # SAMPA104-250
Leverantör:  Sampling Systems
Double Ended Spoons
Double Ended Spoons
Double ended spoon, 250 mm, 1,8/5 ml, Stainless steel
Price
280,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Product Details & Documents
Double ended spoon for taking different sample sizes.
  • Made from stainless steel
  • Ideal for sampling powders and liquids
  • Various sizes available
These double ended spoons are great for taking different sample sizes using the same spoon. The lengths and volume capacities of the spoons vary allowing for flexibility in sampling both powders and liquids.
Specifikationer
  • Capacity:
    1,8/5 ml
  • Description:
    Double ended spoon
  • Length:
    250 mm
  • Material:
    Stainless steel
  • Seq:
    02