Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
tert-butyl (2S)-2-[(1R,2R)-3-[[(1R,2S)-2-hydroxy-1-methyl-2-phenyl-ethyl]amino]-1-methoxy-2-methyl-3-oxo-propyl]pyrrolidine-1-carbox ylate 1 * 1 g
PHBLPBY0192-1G
:
Pharmablock (USA)
tert-butyl (2S)-2-[(1R,2R)-3-[[(1R,2S)-2-hydroxy-1-methyl-2-phenyl-ethyl]amino]-1-methoxy-2-methyl-3-oxo-propyl]pyrrolidine-1-carbox ylate 1 * 1 g
PHBLPBY0192-1G
:
Pharmablock (USA)
Frequently Bought Together
N,N-Dimethylacetamide anhydrous, 99.8 % 1 * 1 L
: Merck
Water ACS
: Thermo Fisher Scientific
Ethyl acetate ≥99%, GPR RECTAPUR®
: VWR Chemicals
tert-Butanol, anhydrous ≥99.5%
: Thermo Fisher Scientific
Ethylene glycol ≥99.0% (by GC), BAKER ANALYZED®, J.T.Baker®
: Avantor
1-Butylboronic acid, Supelco®
: Merck
Ethylene glycol 99.5% for analysis
: Thermo Fisher Scientific
Dichloromethane 99.8% stabilised for HPLC
: Thermo Fisher Scientific
Chloroform ≥99.9% stabilised, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
: Honeywell Chemicals
Methanol ≥99.8% (by GC), Puriss. p.a. Reag. Ph. Eur., ACS, ISO, Riedel-de Haën™
: Honeywell Chemicals
Toluene, low water content ≥99.5% (by GC, corrected for water content), BakerDRY™, J.T.Baker®
: Avantor
Ethyl acetate ≥99.0% (by GC), Laboratory Reagent
: Honeywell Chemicals
Cyclohexane ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
: Honeywell Chemicals