carbon monoxide chloro(hydrido)ruthenium 2-diphenylphosphanyl-N-(2-diphenylphosphanylethyl)ethanamine 1 * 1 g
PHBLPBUC989-1G
carbon monoxide chloro(hydrido)ruthenium 2-diphenylphosphanyl-N-(2-diphenylphosphanylethyl)ethanamine 1 * 1 g
PHBLPBUC989-1G
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Xylene (mixture of isomers) ≥98%, TECHNICAL
: VWR Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
: VWR Chemicals
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals