cis-3,3a,4,5,6,6a-hexahydropyrrolo[3,4-d]oxazol-2-one hydrochloride 1 * 1 g
Catalog # PHBLPBU5766-1-1G
Supplier: Pharmablock (USA)
cis-3,3a,4,5,6,6a-hexahydropyrrolo[3,4-d]oxazol-2-one hydrochloride 1 * 1 g
Catalog # PHBLPBU5766-1-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
N-Methyl-2-pyrrolidone (NMP) 99.5% for analysis
Supplier: Thermo Fisher Scientific
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
ETHYL ACETATE 99.5% FOR SYNTHESIS 1 * 200 L
Supplier: PanReac AppliChem
Methanol bio-based ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Petroleum Ether 40-60C for pesticide analysis 1 * 2,5 L
Supplier: PanReac AppliChem
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
2,2,4-Trimethylpentane ≥99% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Cyclohexane for pesticide residue analysis
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide ≥99.5%, Reagent Grade
Supplier: Honeywell Chemicals
Cyclohexane ≥99.7%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Xylene (mixture of isomers), BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Acetone ≥99.5%, Laboratory Reagent
Supplier: Honeywell Chemicals
Glycerol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 2,5 L
Supplier: PanReac AppliChem