[CIS-3-[[TERT-BUTYL(DIMETHYL)SILYL]OXYME 1 * 1 g
Catalog # PHBLPBU4956-1G
Supplier: Pharmablock (USA)
[CIS-3-[[TERT-BUTYL(DIMETHYL)SILYL]OXYME 1 * 1 g
Catalog # PHBLPBU4956-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Metanol, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Aceton ≥99%, TECHNICAL
Supplier: VWR Chemicals
n-Hexan, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
1-Butanol ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytiskt reagens
Supplier: VWR Chemicals
Etanol 96% (v/v), AnalaR NORMAPUR® analytiskt reagens
Supplier: VWR Chemicals
Xylen ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytiskt reagens
Supplier: VWR Chemicals
Etanol, GPR RECTAPUR®
Supplier: VWR Chemicals
Vatten, LiChrosolv® för kromatografi, for LC-MS, Supelco®
Supplier: Merck
Dietyleter ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Xylen ≥98%, TECHNICAL
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® för HPLC
Supplier: VWR Chemicals