O7-tert-butyl O3-ethyl 1-oxa-2,7-diazaspiro[4.4]non-2-ene-3,7-dicarboxylate 1 * 1 g
Catalog # PHBLPBU1021-1G
Supplier: Pharmablock (USA)
O7-tert-butyl O3-ethyl 1-oxa-2,7-diazaspiro[4.4]non-2-ene-3,7-dicarboxylate 1 * 1 g
Catalog # PHBLPBU1021-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Ethyl acetate anhydrous, 99.8 % 1 * 2 L
Supplier: Merck
n-Heptane, EMPLURA®, Supelco®
Supplier: Merck
Dibutyl ether anhydrous, 99.3 % 1 * 2 L
Supplier: Merck
Benzoic acid methyl ester
Supplier: Merck
Diethyl ether stabilised, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Water for LC-MS 1 * 2,5 L
Supplier: PanReac AppliChem
Water, extra pure, Deionized
Supplier: Thermo Fisher Scientific
Glycerine ≥83.5-88.5% (w/w) in aqueous solution, pure
Supplier: Thermo Fisher Scientific
Acetone ≥99.5% (by GC), liquid USP, BP, Ph. Eur., pure
Supplier: PanReac AppliChem
Ethanol absolute ≥99.8% (by GC), Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetonitrile ≥99.5% ACS
Supplier: Thermo Fisher Scientific
Acetone ≥99.5% ACS
Supplier: VWR Chemicals
Ethanol (untaxed) in water ≥70% (v/v) Ph. Helv., Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol ≥99.8% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
2-Butoxyethanol ≥99%
Supplier: Thermo Fisher Scientific