1-tert-butyl 3-ethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5,6-dihydropyridine-1,3(2H)-dicarboxylate 1 * 1 g
Catalog # PHBLPBEB0946-1G
Supplier: Pharmablock (USA)
1-tert-butyl 3-ethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5,6-dihydropyridine-1,3(2H)-dicarboxylate 1 * 1 g
Catalog # PHBLPBEB0946-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
Glycerine ≥97%, TECHNICAL
Supplier: VWR Chemicals
Pyridine ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Dichloromethane, dehydrated (max. 0.01% H₂O) ≥99% stabilised, GPR RECTAPUR® for synthesis
Supplier: VWR Chemicals
Petroleum spirit, 40…65 °C, anhydrous, TECHNICAL (max. 0.01% aromatic hydrocarbons)
Supplier: VWR Chemicals
Chlorobenzene ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Dichloromethane ≥99.8%, SupraSolv® for GC-MS
Supplier: Merck
n-Hexane ≥98%, SupraSolv® for GC-MS
Supplier: Merck
Diethyl ether stabilised, EMPLURA®, Supelco®
Supplier: Merck
Cyclohexane, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
tert-Butyl methyl ether, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
Ethanol absolute ≥99.5%, TechniSolv®, pure
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Water USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
Dimethyl sulphoxide, extra dry over Molecular Sieve 99.7+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Pyridine, anhydrous ≥99.5%
Supplier: Thermo Fisher Scientific