tert-butyl cis-3-hydroxy-4-(hydroxymethyl)pyrrolidine-1-carboxylate 1 * 1 g
Catalog # PHBLPBAX795-1G
Supplier: Pharmablock (USA)
tert-butyl cis-3-hydroxy-4-(hydroxymethyl)pyrrolidine-1-carboxylate 1 * 1 g
Catalog # PHBLPBAX795-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol, Prepsolv® for preparative chromatography, Supelco®
Supplier: Merck
Cyclohexanone ≥99.0% analytical reagent
Supplier: VWR Chemicals
n-Pentane ≥95%, TECHNICAL
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
2-Propanol, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Glycerine (glycerol) USP, Ph. Eur., BP
Supplier: VWR Chemicals
Trifluoroacetic acid, Sigma-Aldrich®
Supplier: Merck
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
1-Propanol 99+%, extra pure
Supplier: Thermo Fisher Scientific
Ethanol (untaxed) ~96%, Puriss. p.a. ACS, without additive prima fine spirit, F15 (not denatured), Riedel-de Haën™
Supplier: Honeywell Chemicals