Order Entry
Sweden
ContactUsLinkComponent
High Precision Tweezers - Antimagnetic/Anti-acid SS - serrated handles - tips: straight, round, flat, fine. Overall length 120mm 1 * 1 items
Catalog # OUTI321-SA
Supplier:  OUTILS RUBIS
High Precision Tweezers - Antimagnetic/Anti-acid SS - serrated handles - tips: straight, round, flat, fine. Overall length 120mm 1 * 1 items
Catalog # OUTI321-SA
Supplier:  OUTILS RUBIS