5 SPR sensor chips. Biacore Series S compatible. Coating: 80 nm zwitterionic polycarboxylate hydrogel, medium density. Cationic aft er NHS activation. 1 * 5 items
Catalog # MSPPSCBSZC80M-5
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together