Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
CHLORINE STD CL 10µG/G IN 75CST MINERAL 1 * 100 mL
Catalog # PROOVHGCLOIL10-100
Supplier:
LGC Standards PROMOCHEM
Expand image
CHLORINE STD CL 10µG/G IN 75CST MINERAL 1 * 100 mL
Catalog # PROOVHGCLOIL10-100
Supplier:
LGC Standards PROMOCHEM
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Conductivity standards, Certipur® Supelco®
Supplier: Merck
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
Supplier: Merck
UV/Visible standards, Supelco®
Supplier: Merck
Potassium chloride 3 mol/l (3 N) in aqueous solution, Supelco®
Supplier: Merck
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW