Order Entry
Sweden
ContactUsLinkComponent
Capillary Storage Solution, 100mL. Solution necessary for the placement of capillaries between runs. For use with the Fragment Analy zer and ZAG DNA Analyzer systems. 1 * 1 items
Catalog # AGLSGP-440-0100
Supplier:  AGILENT
Capillary Storage Solution, 100mL. Solution necessary for the placement of capillaries between runs. For use with the Fragment Analy zer and ZAG DNA Analyzer systems. 1 * 1 items
Catalog # AGLSGP-440-0100
Supplier:  AGILENT
Frequently Bought Together