Order Entry
Sweden
Orders LinkContactUsLinkComponent
Femto Pulse System
Femto Pulse System
Femto pulse verification kit
Catalog # AGLSFP-OQ
Supplier:   AGILENT
Femto Pulse System
Catalog # AGLSFP-OQ
Supplier:  AGILENT
Femto Pulse System
Femto Pulse System
Femto pulse verification kit
Price
9 537,88 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Pulsed-field capillary electrophoresis system for QC of high molecular weight DNA and RNA samples.
  • Large fragment resolution - separate high molecular weight DNA smears and fragments through 165 kb in about 1,5 hours
  • Femtogram level sensitivity - achieve 10 times higher sensitivity for nucleic acid smears and up to 100 times higher for nucleic acid fragments
  • Simple sample preparation - minimise sample degradation with fewer pipetting steps and stable reagents
  • Powerful analysis software - one intuitive software program automates analysis of diverse sample types
  • Conserve precious sample - unparalleled sensitivity allows you to use less precious sample and achieve superior detection
  • Make workflows more efficient - eliminate pulsed-field gel electrophoresis from long-read NGS library preparation and BAC analysis
  • Desktop PC supplied with a comprehensive 3-year warranty through HP
By using a pulsed-field power supply, large molecular weight DNA fragments can be separated and nucleic acids detected into the femtogram range. Compared to other non-denaturing gel-based analytical instruments, the Femto Pulse system easily achieves 10 times higher sensitivity for nucleic acid smears and up to 100 times higher sensitivity for nucleic acid fragments.
The ability to separate high molecular weight DNA up to 165 kb and detect nucleic acids down to 50 fg/µl input concentration make the Femto Pulse system ideal for long-read NGS QC, gDNA, small RNA, or cfDNA analysis from low concentration samples.
Caution: For research use only. Not for use in diagnostic procedures.
Specifications
  • Description:
    Femto pulse verification kit
Accessories
Product Information
  • Catalog #
    Details
    Availability
    Price
  • 96 well trays for buffer and waste locations, 50 per pack
    96 well trays for buffer and waste locations, 50 per pack
    AGLSP60-20
    For use with the fragment analyser, Femto Pulse, and ZAG DNA analyser systems
    Log in for availability
    1 * 50 Items - 4 120,94 kr
Product Information
  • Catalog #
    Details
    Availability
    Price
  • Large open waste tray/tub
    Large open waste tray/tub
    AGLSP60-22
    For use with the fragment analyser, Femto Pulse, and ZAG DNA analyser systems
    Log in for availability
    1 * 1 Items - 213,88 kr