Order Entry
Sweden
ContactUsLinkComponent
FP US RNA Diluent Marker, 3mL. 15 nt lower marker. For use with the Femto Pulse system and the FP-1201 kit. 1 * 3 mL
Catalog # AGLSFP-8201-0003
Supplier:  AGILENT
FP US RNA Diluent Marker, 3mL. 15 nt lower marker. For use with the Femto Pulse system and the FP-1201 kit. 1 * 3 mL
Catalog # AGLSFP-8201-0003
Supplier:  AGILENT
Frequently Bought Together