Order Entry
Sweden
ContactUsLinkComponent
FP US RNA Ladder, 15 µL. 200-6,000 nt RNA Ladder for use with the Femto Pulse system and the FP-1201 kit 1 * 35 µG
Catalog # AGLSFP-7201-U015
Supplier:  AGILENT
FP US RNA Ladder, 15 µL. 200-6,000 nt RNA Ladder for use with the Femto Pulse system and the FP-1201 kit 1 * 35 µG
Catalog # AGLSFP-7201-U015
Supplier:  AGILENT
Frequently Bought Together