HS NGS DNA Ladder, 100uL. 100-3,000 bp DNA Ladder for use with the Fragment Analyzer systems and the DNF-474 kit 1 * 100 µG
AGLSDNF-396-U100
: AGILENT
HS NGS DNA Ladder, 100uL. 100-3,000 bp DNA Ladder for use with the Fragment Analyzer systems and the DNF-474 kit 1 * 100 µG
AGLSDNF-396-U100
: AGILENT
Frequently Bought Together
VWR® Serological Pipette, Standard Line
: VWR Collection
Fast DNA™ SPIN kit for soil, MP Biomedicals
: MP Biomedicals
Water, PCR grade for PCR
: VWR Chemicals
Decontamination solution, RNase AWAY®
: Thermo Fisher Scientific
Multiwell cell culture plates, Falcon®
: Corning
Kits for Precellys®
: Bertin Instruments
E.Z.N.A.® Plasmid Midi kit
: OMEGA BIO-TEK
HCCA matrix
: BRUKER DALTRONIK
Native barcoding auxiliary kit v14, Oxford Nanopore Technologies
: Oxford Nanopore Technologies
Flow Cell Wash Kit
: Oxford Nanopore Technologies
Ligation Sequencing Kits
: Oxford Nanopore Technologies
Rapid sequencing kits
: Oxford Nanopore Technologies
Direct RNA sequencing kit
: Oxford Nanopore Technologies
sparQ PureMag beads
: Quantabio