Order Entry
Sweden
ContactUsLinkComponent
BF-2000 Blank Solution, 8mL. For wells in which sample is not present necessary for correct capillary alignment. For use with the F ragment Analyzer systems and the DNF-473, DNF-487, and DNF-492 kits. 1 * 8 mL
Catalog # AGLSDNF-302-0008
Supplier:  AGILENT
BF-2000 Blank Solution, 8mL. For wells in which sample is not present necessary for correct capillary alignment. For use with the F ragment Analyzer systems and the DNF-473, DNF-487, and DNF-492 kits. 1 * 8 mL
Catalog # AGLSDNF-302-0008
Supplier:  AGILENT
Frequently Bought Together