SODIUM HYDROXIDE 0.5 MOL/L 1 * 10 L
5613.9010
: Avantor
Frequently Bought Together
Xylene (mixture of isomers) ≥98%, TECHNICAL
: VWR Chemicals
Dimethyl sulphoxide ≥99.5%, BioChemica® cell culture reagent
: PanReac AppliChem
Glycerine ≥98%, GPR RECTAPUR®
: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Petroleum spirit, 60…80 °C, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
2-Propanol ≥99.0% USP, Ph. Eur.
: VWR Chemicals
Pyridine ≥99%, GPR RECTAPUR®
: VWR Chemicals
n-Heptane ≥99.0%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
2,2,4-Trimethylpentane ≥99.5%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Methanol ≥99.8% (by GC), Puriss. p.a. Reag. Ph. Eur., ACS, ISO, Riedel-de Haën™
: Honeywell Chemicals