The antibody was purified from rabbit antiserum by affinity- chromatography using phospho peptide. The antibody against non-phospho peptide was removed by chromatography using corresponding non-phospho peptide 1 * 100 µG
Catalog # AVIVOAAF07835
Leverantör: Aviva Systems Biology