Order Entry
Sweden
Orders LinkContactUsLinkComponent
 

Graphite, powder, TECHNICAL

Plastic drum, 25 kg
Catalog # 24489.460
Supplier:   VWR Chemicals
Graphite, powder, TECHNICAL
Catalog # 24489.460
Supplier:  VWR Chemicals
CAS Number:  7782-42-5
Graphite, powder, TECHNICAL
Graphite, powder, TECHNICAL

Plastic drum, 25 kg

CAS Number:  7782-42-5
Price
4 450,00 kr
Log in for availability
1 * 25 kg
Product Details & Documents
  • CAS Number:7782-42-5
  • Formula:C
  • MDL Number:MFCD00144065
  • Molecular Weight:12.01 g/mol
  • Merck Index:12,04560
  • Melting Pt:3827 °C
  • Density:2.09…2.23 g/cm³ (20 °C)
  • EINECS:231-955-3
Specifications

Specification Test Results

  • Identification:
    Passes test

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    25 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.