0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
Volumetric flasks, USP, BLAUBRAND®, class A, DE-M, amber, 1.000 ml, NS 24/29, Boro 3.3, with glass stopper, USP individual certifica te 1 * 1 items
BRND957471
:
Brand
Volumetric flasks, USP, BLAUBRAND®, class A, DE-M, amber, 1.000 ml, NS 24/29, Boro 3.3, with glass stopper, USP individual certifica te 1 * 1 items
BRND957471
:
Brand