Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
CLOSTRIDIUM AGAR (SHEEP BLOOD 5%) 90 MM 1 * 20 items
Catalog # LIOF11060
Supplier:
LIOFILCHEM
Expand image
CLOSTRIDIUM AGAR (SHEEP BLOOD 5%) 90 MM 1 * 20 items
Catalog # LIOF11060
Supplier:
LIOFILCHEM