ASGR2 MS Standard C13 and N15-labeled recombinant protein (NP_001172) Human Mass Spec Standard 1 * 10 µG
Catalog # ORIGPH313607
Supplier: OriGene
ASGR2 MS Standard C13 and N15-labeled recombinant protein (NP_001172) Human Mass Spec Standard 1 * 10 µG
Catalog # ORIGPH313607
Supplier: OriGene
Frequently Bought Together
Centrifuge tubes, Corning®
Supplier: Corning
VWR®, Centrifuge Tubes, Conical Bottom, with Flat Screw Cap
Supplier: VWR Collection
VWR®, Test Tubes without Rim
Supplier: VWR Collection
VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
VWR® Standard Line Bottles, Narrow Neck, Round, PET, with Screw Caps
Supplier: VWR Collection
Eppendorf® Protein LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
Centrifuge tubes, 15 ml, Corning®
Supplier: Corning
VWR®, Cell Culture Flasks, Non Treated or Treated
Supplier: VWR Collection
PCR tubes and caps, Axygen®
Supplier: Corning
Eppendorf® Safe-Lock Tubes, 0,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Centrifuge Tubes, High Performance, Flat or Plug Cap, PP
Supplier: VWR Collection
Cryoboxes, 133×133 mm
Supplier: RATIOLAB
Serological pipettes, Stripette™
Supplier: Corning
Petri dishes, Star™ Dish
Supplier: PHOENIX BIOMEDICAL
Round-bottom polystyrene tubes, disposable, Falcon®
Supplier: Corning
Centrifuge tubes, sterile, polypropylene, Corning®
Supplier: Corning
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
PLATE P96-1.5H-N. 96 WELL GLASS BOTTOM 1 * 20 ST
Supplier: MARKET SOURCE PART PROCESS