QPCR 96-Well Plates, Non-Skirted 96-Well Non-Skirted PolypropylenePCR Plate for use with Mx3000P / Mx3005P QPCR Systems. Compatible with our Mx3000P / Mx3005P Optical Strip Caps (PN 401425). 25 plates/pack. Agilent offers a range of high quality wel 1 * 25 items
Catalog # AGLS410088
Supplier: AGILENT
Frequently Bought Together