Order Entry
Sweden
ContactUsLinkComponent
Objective HI Plan EPI 20x/0.40 POL - for qualitative and quantitative polarization - Free working distance: 0.9 mm - for use without cover glass - M25 thread 1 * 1 items
Catalog # LEIM11556085
Supplier:  LEICA MICROSYSTEMS
Objective HI Plan EPI 20x/0.40 POL - for qualitative and quantitative polarization - Free working distance: 0.9 mm - for use without cover glass - M25 thread 1 * 1 items
Catalog # LEIM11556085
Supplier:  LEICA MICROSYSTEMS