Order Entry
Sweden
ContactUsLinkComponent
GF Signet Installation Tee Fitting, PVC 80, 4" Pipe ID
GF Signet Installation Tee Fitting, PVC 80, 4" Pipe ID
Catalog # MFLX05620-29
undefined
GF Signet Installation Tee Fitting, PVC 80, 4" Pipe ID
Catalog # MFLX05620-29
About this item
Easily install your flowmeters.
  • Constructed of durable CPVC/PVC 80
PVC Tee Fittings for PVC 80 pipes up to 4" in diameter and CPVC 80 pipes up to 1 1/2" in diameter. Slip on ends (no threads) sensors.
  • Description:
    Installation Tee Fitting, PVC 80, 4" Pipe ID
  • Environmentally Preferable:
  • Warranty:
    1 year part and 2 years labor
  • Connection type:
    4" ID Slip Fit
  • Pk:
    1
  • Fitting material:
    PVC 80