Single-use bioreactor systems, BIOne
Product Details & Documents
Distek, Inc. has developed a bench top scale single-use bioreactor (SUB) for mammalian cell growth, cell therapy, insect cells and other unique applications performed in a stirred tank bioreactor. The gamma irradiated BIOne SUB is engineered with a disposable head plate welded to a triple-layered liner that can be easily inserted into a non sterile bioreactor glass vessel, converting it to a disposable SUB within a matter of seconds.
- Customisable above and below the head plate
- 2 and 5 L working volume
- Optional single-use pH probe
- Increased throughput
- Weldable tubing
- Non invasive DO probe port
- USP class VI materials
- Animal derivative-free and low antioxidant liner material
- Compatible with most manufacturers’ glass vessels and controllers
Upon media addition, the liner expands and moulds to the glass vessel, preserving the geometry of the existing bioreactor. The BIOne SUB’s liner material emulates the large-scale SUBs used in pilot and commercial biopharma production facilities around the world. Performing R&D studies using the most similar material on the market streamlines your tech transfer and can de-risk your process before transferring to pilot scale.
Ordering information: Requires one BIOne starter kit (P/N 2800-0510) for each controller being converted for use with the BIOne SUB. Requires one BIOne motor adapter assembly for each controller being converted for use with the BIOne SUB. Motor adapters are controller motor-specific. For more details please contact your local Avantor sales office.
Documents
Distek BIOne 1250 Bioprocess Control Station Distek BIOne 1250 Controller Distek BIOne SUB
Additional Product Details
Distek BIOne Design of Experiments Upstream Bioprocessing Distek BIOne Effects of Antifoam C Emulsion on kLa Distek BIOne Increasing Oxygen Transfer Using a Microsparger Distek BIOne Recognizing the Potential Impact of Agitation Directionality Distek BIOne Upstream Process Development with a kLa Criterion
Application Notes
TDS Distek BIOne 1250
Datasheet
Specifications
- Beskrivning:BIOne single-use bioreactor with micro sparger, dual impeller and single-use pH probe. Working volume: 2 L
- Seq:16